Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGGGTCCATCTGAAGAGTGCCAAA[A/G]GGAAAAGTAAGACAAAGACATGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605014 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STX11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STX11 - syntaxin 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003764.3 | 207 | Intron | NP_003755.2 | |||
XM_011536213.2 | 207 | UTR 5 | XP_011534515.1 | |||
XM_011536214.2 | 207 | Intron | XP_011534516.1 | |||
XM_011536217.2 | 207 | Intron | XP_011534519.1 | |||
XM_011536218.2 | 207 | Intron | XP_011534520.1 | |||
XM_017011400.1 | 207 | Intron | XP_016866889.1 |