Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAAGCAATACATTCCAATTTCAGA[A/C]ACACTTAAATTCTAGTATTTGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614217 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASCC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASCC3 - activating signal cointegrator 1 complex subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284271.1 | 8085 | Intron | NP_001271200.1 | |||
NM_006828.3 | 8085 | UTR 3 | NP_006819.2 | |||
NM_022091.4 | 8085 | Intron | NP_071374.1 | |||
XM_011535394.2 | 8085 | Intron | XP_011533696.1 | |||
XM_011535395.2 | 8085 | Intron | XP_011533697.1 | |||
XM_011535396.2 | 8085 | Intron | XP_011533698.1 | |||
XM_017010205.1 | 8085 | Intron | XP_016865694.1 | |||
XM_017010206.1 | 8085 | Intron | XP_016865695.1 |