Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACAGTAGCTACCAGATATATTATA[C/T]GGAATAAATGACATTAAGCCACTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 133430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESR1 - estrogen receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000125.3 | Intron | NP_000116.2 | ||||
NM_001122740.1 | Intron | NP_001116212.1 | ||||
NM_001122741.1 | Intron | NP_001116213.1 | ||||
NM_001122742.1 | Intron | NP_001116214.1 | ||||
NM_001291230.1 | Intron | NP_001278159.1 | ||||
NM_001291241.1 | Intron | NP_001278170.1 | ||||
NM_001328100.1 | Intron | NP_001315029.1 | ||||
XM_006715374.3 | Intron | XP_006715437.1 | ||||
XM_006715375.3 | Intron | XP_006715438.1 | ||||
XM_011535543.2 | Intron | XP_011533845.1 | ||||
XM_011535544.2 | Intron | XP_011533846.1 | ||||
XM_011535545.2 | Intron | XP_011533847.1 | ||||
XM_011535547.2 | Intron | XP_011533849.1 | ||||
XM_011535549.2 | Intron | XP_011533851.1 | ||||
XM_017010376.1 | Intron | XP_016865865.1 | ||||
XM_017010377.1 | Intron | XP_016865866.1 | ||||
XM_017010378.1 | Intron | XP_016865867.1 | ||||
XM_017010379.1 | Intron | XP_016865868.1 | ||||
XM_017010380.1 | Intron | XP_016865869.1 | ||||
XM_017010381.1 | Intron | XP_016865870.1 | ||||
XM_017010382.1 | Intron | XP_016865871.1 | ||||
XM_017010383.1 | Intron | XP_016865872.1 |