Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGTAGGTTAAAAATCAGGATG[C/T]CTGGGTGTGAGTGGCAGTGCTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609991 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FNDC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FNDC1 - fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032532.2 | Intron | NP_115921.2 | ||||
XM_011536190.2 | Intron | XP_011534492.1 | ||||
XM_011536191.2 | Intron | XP_011534493.1 |
LOC101929122 - uncharacterized LOC101929122 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |