Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTAGCTTAGCACTTTTGCCTCA[A/G]CTAGTTGTCTCGTTACGGGAAGAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM179B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM179B - family with sequence similarity 179 member B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308120.1 | 752 | Silent Mutation | CAA,CAG | Q,Q 183 | NP_001295049.1 | |
NM_015091.3 | 752 | Silent Mutation | CAA,CAG | Q,Q 183 | NP_055906.2 | |
XM_011536571.1 | 752 | Silent Mutation | CAA,CAG | Q,Q 183 | XP_011534873.1 | |
XM_017021098.1 | 752 | Silent Mutation | CAA,CAG | Q,Q 183 | XP_016876587.1 | |
XM_017021099.1 | 752 | Silent Mutation | CAA,CAG | Q,Q 183 | XP_016876588.1 | |
XM_017021100.1 | 752 | Silent Mutation | CAA,CAG | Q,Q 183 | XP_016876589.1 |
KLHL28 - kelch like family member 28 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308112.1 | 752 | Intron | NP_001295041.1 | |||
NM_017658.4 | 752 | Intron | NP_060128.2 | |||
XM_005267770.3 | 752 | Intron | XP_005267827.1 | |||
XM_006720174.3 | 752 | Intron | XP_006720237.1 | |||
XM_011536847.2 | 752 | Intron | XP_011535149.1 | |||
XM_011536849.2 | 752 | Intron | XP_011535151.1 | |||
XM_017021378.1 | 752 | Intron | XP_016876867.1 |