Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCCTGCAGCTCCCAGGTCCCACG[C/T]TACCTAACTGGACGTTCCTGGCTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605471 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFYVE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFYVE1 - zinc finger FYVE-type containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281734.1 | Intron | NP_001268663.1 | ||||
NM_001281735.1 | Intron | NP_001268664.1 | ||||
NM_021260.3 | Intron | NP_067083.1 | ||||
NM_178441.2 | Intron | NP_848535.1 | ||||
XM_017021373.1 | Intron | XP_016876862.1 |