Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGGGTTGCAATGGTGCCTAAGAT[C/G]CTCTGAAAAAGTGCTATGACTAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AREL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AREL1 - apoptosis resistant E3 ubiquitin protein ligase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039479.1 | 4696 | UTR 3 | NP_001034568.1 | |||
XM_005268244.3 | 4696 | UTR 3 | XP_005268301.1 | |||
XM_006720344.2 | 4696 | UTR 3 | XP_006720407.1 | |||
XM_011537415.2 | 4696 | UTR 3 | XP_011535717.1 | |||
XM_011537416.2 | 4696 | UTR 3 | XP_011535718.1 | |||
XM_011537417.2 | 4696 | UTR 3 | XP_011535719.1 |