Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGCGGGTTACCCTGCAGGTTGAG[A/G]AGGACCAGCCTGGGGCAGGAGGCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601905 MIM: 190195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RABGGTA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RABGGTA - Rab geranylgeranyltransferase alpha subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004581.5 | 2123 | Silent Mutation | CTC,CTT | L,L 537 | NP_004572.3 | |
NM_182836.2 | 2123 | Silent Mutation | CTC,CTT | L,L 537 | NP_878256.1 |
TGM1 - transglutaminase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |