Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAGTTGAGCACGAAATACTTATGT[A/G]TTTGTCCCTGGCTCAAGTCATAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602167 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ESRRB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ESRRB - estrogen related receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004452.3 | Intron | NP_004443.3 | ||||
XM_011536547.2 | Intron | XP_011534849.1 | ||||
XM_011536550.2 | Intron | XP_011534852.1 | ||||
XM_011536553.2 | Intron | XP_011534855.1 | ||||
XM_011536554.2 | Intron | XP_011534856.1 | ||||
XM_017021085.1 | Intron | XP_016876574.1 | ||||
XM_017021086.1 | Intron | XP_016876575.1 |