Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCAGGACAGATAAAGATAATTCA[A/G]TTAGTTGTACTAGAGAGTAACTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605345 MIM: 610211 MIM: 603055 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ALKBH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ALKBH1 - alkB homolog 1, histone H2A dioxygenase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLIRP - SRA stem-loop interacting RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267863.1 | Intron | NP_001254792.1 | ||||
NM_001267864.1 | Intron | NP_001254793.1 | ||||
NM_031210.5 | Intron | NP_112487.1 |
SNW1 - SNW domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |