Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAAATAATTACAATAAAGAAAAA[C/T]AAACCCATATTACAATGATAACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604505 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIP11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIP11 - thyroid hormone receptor interactor 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321851.1 | Intron | NP_001308780.1 | ||||
NM_004239.4 | Intron | NP_004230.2 | ||||
XM_017021787.1 | Intron | XP_016877276.1 | ||||
XM_017021788.1 | Intron | XP_016877277.1 |