Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAAGATAGAGTCAATCTTGGACC[C/T]ATGATGATGACTGGTGGCGTATGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613651 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNORD113-7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SNORD113-7 - small nucleolar RNA, C/D box 113-7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD113-8 - small nucleolar RNA, C/D box 113-8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD113-9 - small nucleolar RNA, C/D box 113-9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD114-1 - small nucleolar RNA, C/D box 114-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD114-2 - small nucleolar RNA, C/D box 114-2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD114-3 - small nucleolar RNA, C/D box 114-3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD114-4 - small nucleolar RNA, C/D box 114-4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD114-5 - small nucleolar RNA, C/D box 114-5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD114-6 - small nucleolar RNA, C/D box 114-6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |