Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGGCAGGGTCCCTGAACCCTGGC[A/G]CTGGGGGAAGTCGCCGACCTGTGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOC3L4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOC3L4 - exocyst complex component 3 like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077594.1 | 368 | Intron | NP_001071062.1 | |||
XM_011537323.2 | 368 | Intron | XP_011535625.1 | |||
XM_011537324.2 | 368 | Intron | XP_011535626.1 | |||
XM_011537325.2 | 368 | UTR 5 | XP_011535627.1 | |||
XM_011537327.2 | 368 | Intron | XP_011535629.1 | |||
XM_011537328.2 | 368 | Intron | XP_011535630.1 | |||
XM_011537329.2 | 368 | Intron | XP_011535631.1 | |||
XM_011537330.2 | 368 | Intron | XP_011535632.1 | |||
XM_011537331.1 | 368 | Intron | XP_011535633.1 | |||
XM_011537332.2 | 368 | Intron | XP_011535634.1 | |||
XM_011537333.2 | 368 | Intron | XP_011535635.1 | |||
XM_011537334.1 | 368 | Intron | XP_011535636.1 |
LOC107984640 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |