Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTTTACACCACACCGCCTCCCT[C/T]AGGTGTCTGGACCATACTGGCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFSD5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MFSD5 - major facilitator superfamily domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170790.1 | 117 | UTR 5 | NP_001164261.1 | |||
NM_032889.4 | 117 | Intron | NP_116278.3 | |||
XM_005269197.1 | 117 | Intron | XP_005269254.1 | |||
XM_005269198.4 | 117 | Intron | XP_005269255.1 |