Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGACGCTTCAAGATCCTGGAGCA[C/T]TGCATGTAAAGAGAAGAGAGAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606371 MIM: 604643 MIM: 605053 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATF7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATF7 - activating transcription factor 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130060.1 | 2937 | UTR 3 | NP_001123532.1 | |||
NM_001206682.1 | 2937 | Intron | NP_001193611.1 | |||
NM_001206683.1 | 2937 | Intron | NP_001193612.1 | |||
NM_006856.2 | 2937 | UTR 3 | NP_006847.1 | |||
XM_005268587.3 | 2937 | UTR 3 | XP_005268644.1 | |||
XM_017018722.1 | 2937 | UTR 3 | XP_016874211.1 |
LOC100652999 - uncharacterized LOC100652999 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPFF - neuropeptide FF-amide peptide precursor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TARBP2 - TARBP2, RISC loading complex RNA binding subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |