Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCATTCCGGGCAGCAATGTGGAG[C/T]GGCCTTGGAAAAAAGGAAAAAGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608730 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD52 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD52 - ankyrin repeat domain 52 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173595.3 | 2982 | Silent Mutation | CCA,CCG | P,P 964 | NP_775866.2 | |
XM_011538197.2 | 2982 | Silent Mutation | CCA,CCG | P,P 903 | XP_011536499.1 | |
XM_017019183.1 | 2982 | Silent Mutation | CCA,CCG | P,P 963 | XP_016874672.1 | |
XM_017019184.1 | 2982 | Intron | XP_016874673.1 |
SLC39A5 - solute carrier family 39 member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |