Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTACTTCTGGTAAATCTACATCCTC[A/G]TCACCACCCATGTTGTTCATCATCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607061 | ||||||||||||||||||||
Literature Links: |
PTGES3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTGES3 - prostaglandin E synthase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282601.1 | 808 | Intron | NP_001269530.1 | |||
NM_001282602.1 | 808 | Silent Mutation | NP_001269531.1 | |||
NM_001282603.1 | 808 | Silent Mutation | NP_001269532.1 | |||
NM_001282604.1 | 808 | Silent Mutation | NP_001269533.1 | |||
NM_001282605.1 | 808 | Intron | NP_001269534.1 | |||
NM_006601.6 | 808 | Silent Mutation | NP_006592.3 | |||
XM_005268576.4 | 808 | Missense Mutation | XP_005268633.1 | |||
XM_006719199.2 | 808 | Missense Mutation | XP_006719262.1 | |||
XM_011537773.2 | 808 | Intron | XP_011536075.1 | |||
XM_011537774.2 | 808 | Intron | XP_011536076.1 | |||
XM_017018716.1 | 808 | Missense Mutation | XP_016874205.1 |