Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAAATAGAAATTTTAGCACATG[A/G]GCCAAGTGTGGTGGCTCATGACTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607664 MIM: 615077 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNS - glucosamine (N-acetyl)-6-sulfatase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D30 - TBC1 domain family member 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015279.1 | Intron | NP_056094.1 | ||||
XM_011538078.2 | Intron | XP_011536380.1 | ||||
XM_017019083.1 | Intron | XP_016874572.1 | ||||
XM_017019084.1 | Intron | XP_016874573.1 | ||||
XM_017019085.1 | Intron | XP_016874574.1 | ||||
XM_017019086.1 | Intron | XP_016874575.1 | ||||
XM_017019087.1 | Intron | XP_016874576.1 |