Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGTGAACCTCCACAATGAGCTGC[A/G]GGGCGACGTCATTCCCCGAGGGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607724 MIM: 610394 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GLIPR1L2 - GLI pathogenesis-related 1 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270396.1 | 241 | Missense Mutation | CAG,CGG | Q,R 65 | NP_001257325.1 | |
NM_152436.2 | 241 | Missense Mutation | CAG,CGG | Q,R 65 | NP_689649.1 | |
XM_011537949.2 | 241 | Intron | XP_011536251.1 | |||
XM_011537950.2 | 241 | Intron | XP_011536252.1 | |||
XM_017018848.1 | 241 | UTR 5 | XP_016874337.1 | |||
XM_017018849.1 | 241 | Intron | XP_016874338.1 |