Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATAAACAACAACAAAGTGACTGG[C/T]ATTTTTTAGCCTTTATTTAATAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615001 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZC3H12C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZC3H12C - zinc finger CCCH-type containing 12C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033390.1 | Intron | NP_203748.1 | ||||
XM_005271715.3 | Intron | XP_005271772.1 | ||||
XM_011543055.2 | Intron | XP_011541357.1 | ||||
XM_017018477.1 | Intron | XP_016873966.1 |