Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCATTCTTCCTTCTTTCTTAACC[A/G]AGGTTACCAGCAGCCACACACATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607784 MIM: 611947 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCG4 - ATP binding cassette subfamily G member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NLRX1 - NLR family member X1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282143.1 | 15 | Intron | NP_001269072.1 | |||
NM_001282144.1 | 15 | Intron | NP_001269073.1 | |||
NM_001282358.1 | 15 | UTR 5 | NP_001269287.1 | |||
NM_024618.3 | 15 | Intron | NP_078894.2 | |||
XM_005271669.1 | 15 | Intron | XP_005271726.1 | |||
XM_005271670.1 | 15 | Intron | XP_005271727.1 | |||
XM_005271671.1 | 15 | Intron | XP_005271728.1 | |||
XM_005271672.1 | 15 | Intron | XP_005271729.1 | |||
XM_006718904.1 | 15 | Intron | XP_006718967.1 | |||
XM_011542980.1 | 15 | UTR 5 | XP_011541282.1 | |||
XM_017018294.1 | 15 | Intron | XP_016873783.1 |