Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTCTCACCTCTGAGCACTGTGAC[A/G]TTCTGGGAGGAGATGGGTGGGGAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188230 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THY1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THY1 - Thy-1 cell surface antigen | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311160.1 | 1397 | Silent Mutation | NP_001298089.1 | |||
NM_001311162.1 | 1397 | Silent Mutation | NP_001298091.1 | |||
NM_006288.4 | 1397 | Silent Mutation | NP_006279.2 |
USP2-AS1 - USP2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |