Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCCAGCCGAGAGCACGATGGCT[A/G]CTGGGGAGCTCGGGGACTTGGGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C11orf70 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C11orf70 - chromosome 11 open reading frame 70 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195005.1 | 69 | Missense Mutation | ACT,GCT | T,A 3 | NP_001181934.1 | |
NM_032930.2 | 69 | Missense Mutation | ACT,GCT | T,A 3 | NP_116319.2 | |
XM_005271713.3 | 69 | Missense Mutation | ACT,GCT | T,A 3 | XP_005271770.1 | |
XM_017018454.1 | 69 | Missense Mutation | ACT,GCT | T,A 3 | XP_016873943.1 |