Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACAATACCAAAAATAAACAAGCT[C/T]AAACTCTTATGATACAATATCTTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MMP27 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP27 - matrix metallopeptidase 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022122.2 | 1508 | Silent Mutation | TTA,TTG | L,L 495 | NP_071405.2 | |
XM_011542948.2 | 1508 | Silent Mutation | TTA,TTG | L,L 452 | XP_011541250.1 | |
XM_011542949.2 | 1508 | Silent Mutation | TTA,TTG | L,L 441 | XP_011541251.1 | |
XM_011542950.2 | 1508 | Silent Mutation | TTA,TTG | L,L 390 | XP_011541252.1 | |
XM_017018120.1 | 1508 | Silent Mutation | TTA,TTG | L,L 494 | XP_016873609.1 |