Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCTTGTTAATTTCACATTCTACA[A/G]AGAAAAGTGCATTTGACTCTTCTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176942 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FER PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FER - FER tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308028.1 | Intron | NP_001294957.1 | ||||
NM_001308031.1 | Intron | NP_001294960.1 | ||||
NM_001308038.1 | Intron | NP_001294967.1 | ||||
NM_005246.3 | Intron | NP_005237.2 | ||||
XM_011543266.1 | Intron | XP_011541568.1 | ||||
XM_011543267.1 | Intron | XP_011541569.1 | ||||
XM_011543269.1 | Intron | XP_011541571.1 | ||||
XM_011543270.2 | Intron | XP_011541572.1 | ||||
XM_011543271.2 | Intron | XP_011541573.1 | ||||
XM_011543272.1 | Intron | XP_011541574.1 | ||||
XM_011543276.2 | Intron | XP_011541578.1 | ||||
XM_011543278.1 | Intron | XP_011541580.1 | ||||
XM_017009229.1 | Intron | XP_016864718.1 | ||||
XM_017009230.1 | Intron | XP_016864719.1 | ||||
XM_017009231.1 | Intron | XP_016864720.1 | ||||
XM_017009232.1 | Intron | XP_016864721.1 | ||||
XM_017009233.1 | Intron | XP_016864722.1 | ||||
XM_017009234.1 | Intron | XP_016864723.1 | ||||
XM_017009235.1 | Intron | XP_016864724.1 |