Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAATTATACTCACATTTGGAGT[C/T]CTGATTCCAAATTTACTATGGAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603943 | ||||||||||||||||||||
Literature Links: |
CDO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDO1 - cysteine dioxygenase type 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323565.1 | 875 | Silent Mutation | AGA,AGG | R,R 207 | NP_001310494.1 | |
NM_001323566.1 | 875 | Silent Mutation | AGA,AGG | R,R 187 | NP_001310495.1 | |
NM_001323567.1 | 875 | Silent Mutation | AGA,AGG | R,R 116 | NP_001310496.1 | |
NM_001801.2 | 875 | Silent Mutation | AGA,AGG | R,R 188 | NP_001792.2 |