Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGTAGCTGTCTGGAGAAGGAGC[A/G]CTGAAGGCAGAGGGAGCAGCCAGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602238 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXOSC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EXOSC2 - exosome component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282708.1 | Intron | NP_001269637.1 | ||||
NM_001282709.1 | Intron | NP_001269638.1 | ||||
NM_014285.6 | Intron | NP_055100.2 | ||||
XM_005272176.2 | Intron | XP_005272233.1 | ||||
XM_006717023.2 | Intron | XP_006717086.1 | ||||
XM_017014558.1 | Intron | XP_016870047.1 |