Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCGGACCTCAAAGTCCTTCCAC[A/G]TCTGCCAGAACGCGCCGGTCGTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612685 MIM: 614770 MIM: 610850 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAMSAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAMSAP3 - calmodulin regulated spectrin associated protein family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6792 - microRNA 6792 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PET100 - PET100 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
XAB2 - XPA binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020196.2 | 2143 | Missense Mutation | ACG,ATG | T,M 706 | NP_064581.2 |