Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGTCCCCGCTGGATTCAAGAAA[T/C]GCTCTGCAGGCAAGGAGTGGCATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609300 | ||||||||||||||||||||
Literature Links: |
CYP17A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CYP17A1 - cytochrome P450 family 17 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000102.3 | 1419 | Missense Mutation | CAT,CGT | H,R 416 | NP_000093.1 |
CYP17A1-AS1 - CYP17A1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC107984264 - transmembrane protein 108-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |