Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGGGATGGGACCTGGGGAAGCCA[C/T]TTCCCACATTTAAATGTTCTATCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179555 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RSU1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RSU1 - Ras suppressor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012425.3 | 2434 | UTR 3 | NP_036557.1 | |||
NM_152724.2 | 2434 | UTR 3 | NP_689937.2 | |||
XM_005252552.3 | 2434 | UTR 3 | XP_005252609.1 |