Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCAGAGCGCCCGCGATCACCTGC[C/G]CCCGGCGGGGCCGCCGGTGCCCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TBC1D12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBC1D12 - TBC1 domain family member 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015188.1 | 1054 | Missense Mutation | CCC,CGC | P,R 290 | NP_056003.1 | |
XM_006717732.3 | 1054 | Missense Mutation | CCC,CGC | P,R 290 | XP_006717795.2 | |
XM_006717733.2 | 1054 | Intron | XP_006717796.1 | |||
XM_011539558.2 | 1054 | Intron | XP_011537860.1 | |||
XM_017015996.1 | 1054 | Missense Mutation | CCC,CGC | P,R 290 | XP_016871485.1 | |
XM_017015997.1 | 1054 | Missense Mutation | CCC,CGC | P,R 290 | XP_016871486.1 | |
XM_017015998.1 | 1054 | Intron | XP_016871487.1 | |||
XM_017015999.1 | 1054 | Intron | XP_016871488.1 |