Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATGTTTCTTAAATAAATGAAT[A/G]GTTTAGGCACAGAGAACAGTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102620 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACTA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACTA2 - actin, alpha 2, smooth muscle, aorta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001141945.2 | Intron | NP_001135417.1 | ||||
NM_001320855.1 | Intron | NP_001307784.1 | ||||
NM_001613.2 | Intron | NP_001604.1 |
ACTA2-AS1 - ACTA2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |