Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCTGTCACTGAGCTACAGAAGT[A/G]TTGATTCCATAAATGTTCCCAGTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603801 MIM: 606116 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMT2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NMT2 - N-myristoyltransferase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308295.1 | 2084 | UTR 3 | NP_001295224.1 | |||
NM_004808.2 | 2084 | UTR 3 | NP_004799.1 | |||
XM_005252642.4 | 2084 | Intron | XP_005252699.1 | |||
XM_006717539.3 | 2084 | UTR 3 | XP_006717602.1 | |||
XM_017016947.1 | 2084 | Intron | XP_016872436.1 | |||
XM_017016948.1 | 2084 | Intron | XP_016872437.1 | |||
XM_017016949.1 | 2084 | Intron | XP_016872438.1 | |||
XM_017016950.1 | 2084 | Intron | XP_016872439.1 | |||
XM_017016951.1 | 2084 | UTR 3 | XP_016872440.1 | |||
XM_017016952.1 | 2084 | Intron | XP_016872441.1 | |||
XM_017016953.1 | 2084 | UTR 3 | XP_016872442.1 |
RPP38 - ribonuclease P/MRP subunit p38 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |