Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCTGGCTGGCTTTCCTTGAGAAT[A/G]TGTATATTTAATCTGTATTTTGTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142600 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HK1 - hexokinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000188.2 | Intron | NP_000179.2 | ||||
NM_001322364.1 | Intron | NP_001309293.1 | ||||
NM_001322365.1 | Intron | NP_001309294.1 | ||||
NM_001322366.1 | Intron | NP_001309295.1 | ||||
NM_001322367.1 | Intron | NP_001309296.1 | ||||
NM_033496.2 | Intron | NP_277031.1 | ||||
NM_033497.2 | Intron | NP_277032.1 | ||||
NM_033498.2 | Intron | NP_277033.1 | ||||
NM_033500.2 | Intron | NP_277035.2 | ||||
XM_005269737.1 | Intron | XP_005269794.1 | ||||
XM_011539732.1 | Intron | XP_011538034.1 |