Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGAGGGAACACTGTTCTTAGCAG[A/C]AGGATATTTGAGCTCTAATTTCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MALRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MALRD1 - MAM and LDL receptor class A domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142308.2 | Intron | NP_001135780.2 | ||||
XM_011519453.2 | Intron | XP_011517755.1 | ||||
XM_011519454.1 | Intron | XP_011517756.1 | ||||
XM_011519455.2 | Intron | XP_011517757.1 | ||||
XM_017016182.1 | Intron | XP_016871671.1 | ||||
XM_017016183.1 | Intron | XP_016871672.1 | ||||
XM_017016184.1 | Intron | XP_016871673.1 | ||||
XM_017016185.1 | Intron | XP_016871674.1 | ||||
XM_017016186.1 | Intron | XP_016871675.1 |