Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTCATAAGTTGTCCTGGAACAG[A/G]TGCCCCACTGGCTGCTTTAGAACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613923 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPM - lipase family member M | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128215.1 | Intron | NP_001121687.1 | ||||
XM_011539748.2 | Intron | XP_011538050.1 | ||||
XM_011539751.2 | Intron | XP_011538053.1 | ||||
XM_011539752.2 | Intron | XP_011538054.1 | ||||
XM_011539753.2 | Intron | XP_011538055.1 |