Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCTCTTTTAGCTGCAGTTTGAGC[A/T]TCAGGCAAATCTTCTTCCTCTTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602856 | ||||||||||||||||||||
Literature Links: |
RGS10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RGS10 - regulator of G-protein signaling 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005339.1 | 573 | Missense Mutation | GAA,GAT | E,D 167 | NP_001005339.1 | |
NM_002925.3 | 573 | Missense Mutation | GAA,GAT | E,D 153 | NP_002916.1 | |
XM_017016492.1 | 573 | Missense Mutation | GAA,GAT | E,D 105 | XP_016871981.1 | |
XM_017016493.1 | 573 | Missense Mutation | GAA,GAT | E,D 119 | XP_016871982.1 |