Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGCTTCAGCCACTGGACAAAGC[A/G]CTTCATTTCTAGGTAGCTGCTGTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609682 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCLRE1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCLRE1A - DNA cross-link repair 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271816.1 | 3788 | Missense Mutation | CGC,TGC | R,C 1002 | NP_001258745.1 | |
NM_014881.4 | 3788 | Missense Mutation | CGC,TGC | R,C 1002 | NP_055696.3 | |
XM_006718090.1 | 3788 | Missense Mutation | CGC,TGC | R,C 1002 | XP_006718153.1 | |
XM_011540429.1 | 3788 | Missense Mutation | CGC,TGC | R,C 1002 | XP_011538731.1 |