Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGGTTTATAGGCCCAAAAGGTC[A/G]ACGAATTTTCCTCATTCTCAGAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC35G1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC35G1 - solute carrier family 35 member G1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134658.1 | 412 | Missense Mutation | CAA,CGA | Q,R 129 | NP_001128130.1 | |
NM_153226.2 | 412 | Missense Mutation | CAA,CGA | Q,R 128 | NP_694958.1 | |
XM_011539371.2 | 412 | Missense Mutation | CAA,CGA | Q,R 112 | XP_011537673.1 | |
XM_011539373.2 | 412 | Intron | XP_011537675.1 |