Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGCTGCTGGAGAAGGTAGTAAA[A/G]CAGGGCTGTCACCAGCTAATTGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611448 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BMS1 - BMS1, ribosome biogenesis factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014753.3 | 1734 | Missense Mutation | ACA,GCA | T,A 549 | NP_055568.3 | |
XM_005271846.3 | 1734 | Missense Mutation | ACA,GCA | T,A 549 | XP_005271903.1 | |
XM_005271848.3 | 1734 | Missense Mutation | ACA,GCA | T,A 549 | XP_005271905.1 | |
XM_005271849.3 | 1734 | Missense Mutation | ACA,GCA | T,A 549 | XP_005271906.1 | |
XM_011540402.2 | 1734 | Missense Mutation | ACA,GCA | T,A 549 | XP_011538704.1 | |
XM_011540403.2 | 1734 | Missense Mutation | ACA,GCA | T,A 138 | XP_011538705.1 |