Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAACGTGGGCTCAGGTGTACTCG[A/G]TTTTGCGGATATAATTGGAGGGAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611282 | ||||||||||||||||||||
Literature Links: |
DNMBP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNMBP - dynamin binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318326.1 | 4713 | Missense Mutation | ACC,ATC | T,I 1206 | NP_001305255.1 | |
NM_001318327.1 | 4713 | Missense Mutation | ACC,ATC | T,I 862 | NP_001305256.1 | |
NM_015221.3 | 4713 | Missense Mutation | ACC,ATC | T,I 1574 | NP_056036.1 | |
XM_006717735.2 | 4713 | Missense Mutation | ACC,ATC | T,I 1531 | XP_006717798.1 | |
XM_011539559.2 | 4713 | Missense Mutation | ACC,ATC | T,I 1574 | XP_011537861.1 |