Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGGTTTTCTGTCTCATAGCTAT[C/T]GGAATGAACATGATAAACCTGTCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609851 | ||||||||||||||||||||
Literature Links: |
IPMK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IPMK - inositol polyphosphate multikinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152230.4 | 555 | Intron | NP_689416.1 | |||
XM_011539565.2 | 555 | Intron | XP_011537867.1 | |||
XM_017016013.1 | 555 | Missense Mutation | AAT,GAT | N,D 101 | XP_016871502.1 |