Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCGCACTGTCTTTGTATCTGAGC[A/G]GGAGCTGGACTGGGCCAAGGTGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615570 | ||||||||||||||||||||
Literature Links: |
SFXN2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SFXN2 - sideroflexin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178858.4 | 336 | Missense Mutation | CAG,CGG | Q,R 41 | NP_849189.1 | |
XM_011539261.2 | 336 | Missense Mutation | CAG,CGG | Q,R 50 | XP_011537563.1 | |
XM_011539262.2 | 336 | Missense Mutation | CAG,CGG | Q,R 41 | XP_011537564.1 | |
XM_011539263.2 | 336 | UTR 5 | XP_011537565.1 | |||
XM_011539264.2 | 336 | UTR 5 | XP_011537566.1 | |||
XM_017015663.1 | 336 | Missense Mutation | CAG,CGG | Q,R 59 | XP_016871152.1 |