Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAAGCAGATGGCAGCAAGGTTT[A/G]ACAGAGAGAAAGAAGGCCAAACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613847 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TCTN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TCTN3 - tectonic family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143973.1 | Intron | NP_001137445.1 | ||||
NM_015631.5 | Intron | NP_056446.4 | ||||
XM_005269690.1 | Intron | XP_005269747.1 | ||||
XM_011539627.1 | Intron | XP_011537929.1 | ||||
XM_011539628.1 | Intron | XP_011537930.1 |