Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGGTAGACCTCATAGGTTGTATA[A/T]GCTGCTAAGTCTTGTCCAAGGGTTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
A1CF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
A1CF - APOBEC1 complementation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198818.1 | 1973 | Silent Mutation | GCA,GCT | A,A 564 | NP_001185747.1 | |
NM_001198819.1 | 1973 | Silent Mutation | GCA,GCT | A,A 580 | NP_001185748.1 | |
NM_001198820.1 | 1973 | Silent Mutation | GCA,GCT | A,A 572 | NP_001185749.1 | |
NM_014576.3 | 1973 | Silent Mutation | GCA,GCT | A,A 564 | NP_055391.2 | |
NM_138932.2 | 1973 | Silent Mutation | GCA,GCT | A,A 572 | NP_620310.1 | |
NM_138933.2 | 1973 | Silent Mutation | GCA,GCT | A,A 572 | NP_620311.1 | |
XM_005269718.2 | 1973 | Silent Mutation | GCA,GCT | A,A 572 | XP_005269775.1 | |
XM_005269720.3 | 1973 | Silent Mutation | GCA,GCT | A,A 572 | XP_005269777.1 | |
XM_011539729.2 | 1973 | Silent Mutation | GCA,GCT | A,A 580 | XP_011538031.1 | |
XM_011539730.2 | 1973 | Silent Mutation | GCA,GCT | A,A 525 | XP_011538032.1 | |
XM_017016160.1 | 1973 | Silent Mutation | GCA,GCT | A,A 564 | XP_016871649.1 |