Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGACAATACCAGCTCTCTCAATG[C/T]TTTTCTGCTGCAGTGTATACAAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613189 MIM: 606160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR107 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR107 - microRNA 107 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PANK1 - pantothenate kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138316.3 | Intron | NP_612189.2 | ||||
NM_148977.2 | Intron | NP_683878.1 | ||||
NM_148978.2 | Intron | NP_683879.1 | ||||
XM_017016333.1 | Intron | XP_016871822.1 | ||||
XM_017016334.1 | Intron | XP_016871823.1 | ||||
XM_017016335.1 | Intron | XP_016871824.1 | ||||
XM_017016336.1 | Intron | XP_016871825.1 | ||||
XM_017016337.1 | Intron | XP_016871826.1 | ||||
XM_017016338.1 | Intron | XP_016871827.1 |