Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTAACCATGGGCTGTTTTCCA[C/T]GGGGCTAGAACCCCTCTTCACGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C10orf128 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C10orf128 - chromosome 10 open reading frame 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010863.2 | 616 | UTR 3 | NP_001010863.1 | |||
NM_001288740.1 | 616 | UTR 3 | NP_001275669.1 | |||
NM_001288741.1 | 616 | UTR 3 | NP_001275670.1 | |||
NM_001288742.1 | 616 | UTR 3 | NP_001275671.1 | |||
NM_001288743.1 | 616 | UTR 3 | NP_001275672.1 | |||
XM_011539378.2 | 616 | Intron | XP_011537680.1 | |||
XM_011539379.1 | 616 | UTR 3 | XP_011537681.1 | |||
XM_011539380.1 | 616 | UTR 3 | XP_011537682.1 | |||
XM_011539382.1 | 616 | Intron | XP_011537684.1 | |||
XM_011539383.2 | 616 | UTR 3 | XP_011537685.1 | |||
XM_011539384.1 | 616 | UTR 3 | XP_011537686.1 | |||
XM_011539385.1 | 616 | UTR 3 | XP_011537687.1 | |||
XM_017015792.1 | 616 | Intron | XP_016871281.1 | |||
XM_017015793.1 | 616 | Intron | XP_016871282.1 | |||
XM_017015794.1 | 616 | Intron | XP_016871283.1 | |||
XM_017015795.1 | 616 | Intron | XP_016871284.1 | |||
XM_017015796.1 | 616 | Intron | XP_016871285.1 | |||
XM_017015797.1 | 616 | Intron | XP_016871286.1 |
FAM170B-AS1 - FAM170B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |