Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAACCCTGCACACCCACCATGGAT[A/G]TCTTCAAGAAGGGCTTCTCCATCGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608925 MIM: 602998 | ||||||||||||||||||||
Literature Links: |
ADIRF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADIRF - adipogenesis regulatory factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MMRN2 - multimerin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024756.2 | 174 | Intron | NP_079032.2 | |||
XM_005270153.1 | 174 | Intron | XP_005270210.1 | |||
XM_006717970.3 | 174 | Intron | XP_006718033.1 |
SNCG - synuclein gamma | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003087.2 | 174 | Missense Mutation | ATC,GTC | I,V 3 | NP_003078.2 | |
XM_011540105.2 | 174 | Missense Mutation | ATC,GTC | I,V 3 | XP_011538407.1 |