Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACGCAATTCCTCCACAGTACAC[A/G]GTACAATTTTTTGGACTTGACTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIAL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TIAL1 - TIA1 cytotoxic granule-associated RNA binding protein-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033925.1 | 1244 | Silent Mutation | ACC,ACT | T,T 223 | NP_001029097.1 | |
NM_001323964.1 | 1244 | Silent Mutation | ACC,ACT | T,T 83 | NP_001310893.1 | |
NM_001323965.1 | 1244 | Silent Mutation | ACC,ACT | T,T 83 | NP_001310894.1 | |
NM_001323967.1 | 1244 | Silent Mutation | ACC,ACT | T,T 83 | NP_001310896.1 | |
NM_001323968.1 | 1244 | Silent Mutation | ACC,ACT | T,T 184 | NP_001310897.1 | |
NM_001323969.1 | 1244 | Silent Mutation | ACC,ACT | T,T 167 | NP_001310898.1 | |
NM_001323970.1 | 1244 | Silent Mutation | ACC,ACT | T,T 184 | NP_001310899.1 | |
NM_003252.3 | 1244 | Silent Mutation | ACC,ACT | T,T 206 | NP_003243.1 | |
XM_017016605.1 | 1244 | Intron | XP_016872094.1 | |||
XM_017016606.1 | 1244 | Silent Mutation | ACC,ACT | T,T 83 | XP_016872095.1 |