Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCCCATGCAAGTTTCTTCTTTTTT[C/T]GGGCAGCCTGTGAATTTTCAACCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM204A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM204A - family with sequence similarity 204 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134672.1 | 1070 | Missense Mutation | CAA,CGA | Q,R 209 | NP_001128144.1 | |
NM_022063.2 | 1070 | Missense Mutation | CAA,CGA | Q,R 209 | NP_071346.1 | |
XM_005270024.1 | 1070 | Missense Mutation | CAA,CGA | Q,R 216 | XP_005270081.1 |